Assay Details
| Gene Symbol: CHGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164005.2 | NP_001157477.2 | 2 - 3 | 299 | 62 | |
| GenBank mRNA | AK347690.1 | - | 2 - 3 | 282 | 62 | |
| DB783674.1 | - | 2 - 3 | 281 | 62 | ||
| FS658372.1 | - | 2 - 3 | 282 | 62 | ||
| M20926.1 | - | 2 - 3 | 87 | 62 | ||
More Information
Set Membership: |
|

CHGA
Genomic Map