Assay Details
| Gene Symbol: FGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124010.5 | XP_003124058.1 | 892 | 64 | ||
| XM_013987617.2 | XP_013843071.1 | 981 | 64 | |||
| XM_013987618.2 | XP_013843072.1 | 738 | 64 | |||
| XM_021084948.1 | XP_020940607.1 | 376 | 64 | |||
| GenBank mRNA | X60317.1 | - | 290 | 64 | ||
More Information
Set Membership: |
|

FGF1
Genomic Map