Assay Details
| Gene Symbol: LBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128435.1 | NP_001121907.1 | 10 | 89 | ||
| GenBank mRNA | AF268388.1 | - | 3 | 256 | 89 | |
| AK345857.1 | - | 10 | 1052 | 89 | ||
| EF626694.1 | - | 10 | 89 | |||
More Information
Set Membership: |
|

LBP
Genomic Map