Assay Details
| Gene Symbol: RPL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131509.5 | XP_003131557.1 | 219 | 68 | ||
| GenBank mRNA | AF435591.1 | - | 1 | 52 | 68 | |
| AK234321.1 | - | 2 | 107 | 68 | ||
| AK346103.1 | - | 2 | 108 | 68 | ||
| AK392766.1 | - | 2 | 106 | 68 | ||
| AY610419.1 | - | 1 | 90 | 68 | ||
More Information
Set Membership: |
|

RPL19
Genomic Map