Assay Details
| Gene Symbol: UBE2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661722.3 | XP_005661779.2 | 722 | 139 | ||
| GenBank mRNA | AK350560.1 | - | 5 | 728 | 139 | |
| AY148222.1 | - | 4 | 254 | 139 | ||
More Information
Set Membership: |
|

UBE2D2
Genomic Map