Assay Details
| Gene Symbol: ATP5MG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244137.1 | NP_001231066.1 | 3 | 514 | 80 | |
| GenBank mRNA | AK234004.1 | - | 3 | 290 | 80 | |
| AK238581.1 | - | 3 | 273 | 80 | ||
| AK343840.1 | - | 3 | 262 | 80 | ||
| AK343890.1 | - | 3 | 276 | 80 | ||
| AY280702.1 | - | 1 | 44 | 80 | ||
| CN162223.1 | - | 3 | 514 | 80 | ||
More Information
Set Membership: |
|

ATP5MG
Genomic Map