Assay Details
| Gene Symbol: C1QC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244286.1 | NP_001231215.1 | 3 | 658 | 66 | |
| XM_021093594.1 | XP_020949253.1 | 605 | 66 | |||
| GenBank mRNA | AK237170.1 | - | 3 | 626 | 66 | |
| AK237244.1 | - | 3 | 807 | 66 | ||
| AK237332.1 | - | 3 | 627 | 66 | ||
| AK394392.1 | - | 3 | 627 | 66 | ||
| AK397482.1 | - | 3 | 654 | 66 | ||
| AK397557.1 | - | 3 | 654 | 66 | ||
| AK397596.1 | - | 3 | 810 | 66 | ||
| AY349425.1 | - | 3 | 552 | 66 | ||
| CK461627.1 | - | 1 | 501 | 66 | ||
More Information
Set Membership: |
|

C1QC
Genomic Map