Assay Details
| Gene Symbol: HMOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244412.1 | NP_001231341.1 | 4 | 349 | 75 | |
| XM_013995672.2 | XP_013851126.1 | 415 | 75 | |||
| XM_013995673.2 | XP_013851127.1 | 380 | 75 | |||
| XM_021085820.1 | XP_020941479.1 | 278 | 75 | |||
| GenBank mRNA | AK392679.1 | - | 4 | 335 | 75 | |
| AY421755.1 | - | 1 | 44 | 75 | ||
| EW107871.2 | - | 4 | 355 | 75 | ||
More Information
Set Membership: |
|

HMOX2
Genomic Map