Assay Details
| Gene Symbol: RUNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095591.1 | XP_020951250.1 | 877 | 89 | ||
| XM_021095592.1 | XP_020951251.1 | 1165 | 89 | |||
| GenBank mRNA | AY515225.1 | - | 1 | 49 | 89 | |
More Information
Set Membership: |
|

RUNX3
Genomic Map