Assay Details
| Gene Symbol: UPF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135340.4 | XP_003135388.1 | 1290 | 117 | ||
| XM_003135341.5 | XP_003135389.1 | 1331 | 117 | |||
| GenBank mRNA | AY553923.1 | - | 2 | 254 | 117 | |
More Information
Set Membership: |
|

UPF3B
Genomic Map