Assay Details
| Gene Symbol: GC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356971.4 | XP_003357019.1 | 1498 | 58 | ||
| GenBank mRNA | AY609552.1 | - | 11 | 1393 | 58 | |
| AY710291.1 | - | 5 | 548 | 58 | ||
More Information
Set Membership: |
|

GC
Genomic Map