Assay Details
| Gene Symbol: RPL13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244068.1 | NP_001230997.1 | 4 | 251 | 77 | |
| NM_001244069.1 | NP_001230998.1 | 4 | 248 | 77 | ||
| XM_005664723.3 | XP_005664780.1 | 145 | 77 | |||
| XM_013998640.2 | XP_013854094.1 | 1201 | 77 | |||
| XM_013998641.2 | XP_013854095.1 | 1150 | 77 | |||
| XM_013998642.2 | XP_013854096.1 | 1191 | 77 | |||
| XM_013998643.2 | XP_013854097.1 | 1345 | 77 | |||
| XM_021093617.1 | XP_020949276.1 | 1205 | 77 | |||
| XM_021093619.1 | XP_020949278.1 | 1179 | 77 | |||
| GenBank mRNA | AK231513.1 | - | 3 | 173 | 77 | |
| AK232071.1 | - | 4 | 254 | 77 | ||
| AK233946.1 | - | 2 | 78 | |||
| AK344032.1 | - | 3 | 159 | 77 | ||
| AK392037.1 | - | 4 | 249 | 77 | ||
| AK397300.1 | - | 2 | 124 | 77 | ||
| AK398989.1 | - | 3 | 154 | 77 | ||
| DQ178127.1 | - | 1 | 72 | 77 | ||
| EB414714.1 | - | 4 | 235 | 77 | ||
| FS667913.1 | - | 4 | 253 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005664723.3 transcripts.
Set Membership: |
|

RPL13A
Genomic Map