Assay Details
| Gene Symbol: PCNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291925.1 | NP_001278854.1 | 4 | 563 | 112 | |
| GenBank mRNA | AK398386.1 | - | 4 | 681 | 112 | |
| AY610109.1 | - | 4 | 692 | 112 | ||
| DQ473295.1 | - | 4 | 349 | 112 | ||
| GQ913657.1 | - | 4 | 563 | 112 | ||
More Information
Set Membership: |
|

PCNA
Genomic Map