Assay Details
| Gene Symbol: CYP27A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243304.1 | NP_001230233.1 | 7 - 8 | 1933 | 77 | |
| GenBank mRNA | AK232936.1 | - | 7 - 8 | 1933 | 77 | |
| AK392950.1 | - | 7 - 8 | 1052 | 77 | ||
| AK400072.1 | - | 7 - 8 | 1346 | 77 | ||
| EF625352.1 | - | 1 - 2 | 54 | 77 | ||
More Information
Set Membership: |
|

CYP27A1
Genomic Map