Assay Details
| Gene Symbol: ATP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001258.1 | NP_001001258.1 | 7 | 852 | 65 | |
| XM_005653839.3 | XP_005653896.1 | 873 | 65 | |||
| GenBank mRNA | M35497.1 | - | 7 | 852 | 65 | |
More Information
Set Membership: |
|

ATP4B
Genomic Map