Assay Details
| Gene Symbol: AKR1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001539.2 | NP_001001539.1 | 8 - 9 | 851 | 80 | |
| XM_021078497.1 | XP_020934156.1 | 1157 | 80 | |||
| XM_021078498.1 | XP_020934157.1 | 1185 | 80 | |||
| GenBank mRNA | AK234773.1 | - | 8 - 9 | 936 | 80 | |
| AK347681.1 | - | 8 - 9 | 888 | 80 | ||
| AK396909.1 | - | 10 - 11 | 1068 | 80 | ||
| AK398724.1 | - | 8 - 9 | 922 | 80 | ||
| AK400749.1 | - | 8 - 9 | 890 | 80 | ||
| L14950.1 | - | 8 - 9 | 854 | 80 | ||
| U46065.1 | - | 8 - 9 | 854 | 80 | ||
More Information
Set Membership: |
|

AKR1B1
Genomic Map