Assay Details
| Gene Symbol: CCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001619.1 | NP_001001619.1 | 2 | 1348 | 76 | |
| GenBank mRNA | AB119271.1 | - | 2 | 1348 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001619.1 | NP_001001619.1 | 2 | 1348 | 76 | |
| GenBank mRNA | AB119271.1 | - | 2 | 1348 | 76 | |
Set Membership: |
|