Assay Details
| Gene Symbol: CCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001624.1 | NP_001001624.1 | 3 | 1149 | 73 | |
| GenBank mRNA | AB119263.1 | - | 3 | 1149 | 73 | |
| AK239581.1 | - | 3 | 1190 | 73 | ||
| AK351895.1 | - | 3 | 1146 | 73 | ||
| AK398376.1 | - | 3 | 1156 | 73 | ||
More Information
Set Membership: |
|

CCR9
Genomic Map