Assay Details
| Gene Symbol: RPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001638.1 | NP_001001638.1 | 4 | 454 | 72 | |
| GenBank mRNA | AK233933.1 | - | 5 | 500 | 72 | |
| AK348796.1 | - | 5 | 471 | 72 | ||
| AK389481.1 | - | 3 | 1035 | 72 | ||
| AK391988.1 | - | 5 | 478 | 72 | ||
| AY547313.1 | - | 4 | 454 | 72 | ||
| AY550042.1 | - | 4 | 406 | 72 | ||
| AY610406.1 | - | 5 | 479 | 72 | ||
More Information
Set Membership: |
|

RPL11
Genomic Map