Assay Details
| Gene Symbol: IFNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003923.1 | NP_001003923.1 | 1 | 454 | 79 | |
| GenBank mRNA | AY687281.1 | - | 1 | 454 | 79 | |
| EF104599.1 | - | 1 | 454 | 79 | ||
| EU744562.1 | - | 1 | 388 | 79 | ||
| GQ415073.1 | - | 1 | 454 | 79 | ||
| KF414741.1 | - | 1 | 454 | 79 | ||
| KJ147517.1 | - | 1 | 454 | 79 | ||
More Information
Set Membership: |
|

IFNB1
Genomic Map