Assay Details
| Gene Symbol: P2RY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006591.1 | NP_001006591.1 | 2 | 1149 | 78 | |
| XM_021102250.1 | XP_020957909.1 | 1451 | 78 | |||
| XM_021102251.1 | XP_020957910.1 | 1675 | 78 | |||
| XM_021102252.1 | XP_020957911.1 | 2509 | 78 | |||
| GenBank mRNA | AK240593.1 | - | 1 | 563 | 78 | |
| AY620400.1 | - | 2 | 1149 | 78 | ||
More Information
Set Membership: |
|

P2RY2
Genomic Map