Assay Details
| Gene Symbol: MYLPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006592.1 | NP_001006592.1 | 6 | 448 | 67 | |
| XM_021085892.1 | XP_020941551.1 | 523 | 67 | |||
| GenBank mRNA | AK347109.1 | - | 6 | 481 | 67 | |
| AY754870.1 | - | 6 | 448 | 67 | ||
| DQ629157.1 | - | 6 | 459 | 67 | ||
More Information
Set Membership: |
|

MYLPF
Genomic Map