Assay Details
| Gene Symbol: AGRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011693.1 | NP_001011693.1 | 3 | 355 | 63 | |
| XM_021093546.1 | XP_020949205.1 | 1249 | 63 | |||
| GenBank mRNA | AF220531.1 | - | 3 | 318 | 63 | |
More Information
Set Membership: |
|

AGRP
Genomic Map