Assay Details
| Gene Symbol: HAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014428.1 | NP_001014428.1 | 1 | 746 | 120 | |
| XM_005672594.3 | XP_005672651.2 | 806 | 123 | |||
| GenBank mRNA | AY940738.1 | - | 1 | 746 | 120 | |
More Information
Set Membership: |
|

HAND1
Genomic Map