Assay Details
| Gene Symbol: DSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035535.1 | NP_001030612.1 | 15 | 3050 | 87 | |
| GenBank mRNA | AY677210.1 | - | 15 | 3050 | 87 | |
More Information
Set Membership: |
|

DSG1
Genomic Map