Assay Details
| Gene Symbol: SRSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038007.1 | NP_001033096.1 | 3 - 4 | 648 | 118 | |
| GenBank mRNA | DQ098950.1 | - | 3 - 4 | 648 | 118 | |
More Information
Set Membership: |
|

SRSF1
Genomic Map