Assay Details
| Gene Symbol: RPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044543.1 | NP_001038008.1 | 4 | 456 | 84 | |
| GenBank mRNA | AK240170.1 | - | 4 | 431 | 84 | |
| AK343531.1 | - | 1 | 1318 | 84 | ||
| AK351230.1 | - | 5 | 326 | 84 | ||
| AY610342.1 | - | 4 | 310 | 84 | ||
| AY735455.1 | - | 4 | 456 | 84 | ||
More Information
Set Membership: |
|

RPL10
Genomic Map