Assay Details
| Gene Symbol: ARL6IP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048073.1 | NP_001041538.1 | 2 | 249 | 71 | |
| GenBank mRNA | AK234908.1 | - | 2 | 342 | 71 | |
| AK239471.1 | - | 2 | 263 | 71 | ||
| AK351446.1 | - | 2 | 250 | 71 | ||
| AK396096.1 | - | 2 | 374 | 71 | ||
| AK396114.1 | - | 2 | 264 | 71 | ||
| AK400807.1 | - | 2 | 494 | 71 | ||
| AY610408.1 | - | 2 | 242 | 71 | ||
| AY864608.1 | - | 2 | 249 | 71 | ||
More Information
Set Membership: |
|

ARL6IP5
Genomic Map