Assay Details
| Gene Symbol: SMPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078687.2 | NP_001072155.1 | 3 - 4 | 243 | 122 | |
| GenBank mRNA | AY610443.1 | - | 3 - 4 | 297 | 122 | |
| DQ104414.1 | - | 3 - 4 | 334 | 122 | ||
| DQ845243.1 | - | 3 - 4 | 244 | 122 | ||
More Information
Set Membership: |
|

SMPX
Genomic Map