Assay Details
| Gene Symbol: PAQR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213740.1 | NP_998905.1 | 2 | 1047 | 109 | |
| GenBank mRNA | AF313615.1 | - | 2 | 1047 | 109 | |
| AK391615.1 | - | 2 | 1061 | 109 | ||
More Information
Set Membership: |
|

PAQR8
Genomic Map