Assay Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213755.2 | NP_998920.2 | 7 | 853 | 73 | |
| GenBank mRNA | AB530157.1 | - | 7 | 757 | 73 | |
| AK235017.1 | - | 7 | 877 | 73 | ||
| AK236141.1 | - | 7 | 879 | 73 | ||
| AK236691.1 | - | 7 | 879 | 73 | ||
| AK389567.1 | - | 7 | 884 | 73 | ||
| AK395342.1 | - | 7 | 884 | 73 | ||
| AK395662.1 | - | 7 | 877 | 73 | ||
| AK398790.1 | - | 7 | 880 | 73 | ||
| AK400703.1 | - | 8 | 993 | 73 | ||
| AY368628.1 | - | 7 | 831 | 73 | ||
| AY800265.1 | - | 7 | 853 | 73 | ||
| U53020.1 | - | 7 | 879 | 73 | ||
More Information
Set Membership: |
|

STAR
Genomic Map