Assay Details
| Gene Symbol: SULT1A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213765.1 | NP_998930.1 | 8 | 952 | 64 | |
| GenBank mRNA | AK351188.1 | - | 8 | 965 | 64 | |
| AK351208.1 | - | 8 | 1003 | 64 | ||
| AK393906.1 | - | 8 | 983 | 64 | ||
| AK394201.1 | - | 8 | 1007 | 64 | ||
| AK398946.1 | - | 8 | 989 | 64 | ||
| AK400970.1 | - | 8 | 965 | 64 | ||
| AY193893.1 | - | 8 | 952 | 64 | ||
More Information
Set Membership: |
|

SULT1A3
Genomic Map