Assay Details
| Gene Symbol: IFNAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213772.1 | NP_998937.1 | 11 | 1555 | 107 | |
| XM_021068507.1 | XP_020924166.1 | 1697 | 107 | |||
| GenBank mRNA | AB116561.1 | - | 11 | 1555 | 107 | |
| AK347850.1 | - | 11 | 1692 | 107 | ||
| AK390686.1 | - | 3 | 364 | 107 | ||
| AK396446.1 | - | 11 | 1691 | 107 | ||
| AK400959.1 | - | 2 | 225 | 107 | ||
More Information
Set Membership: |
|

IFNAR1
Genomic Map