Assay Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213776.1 | NP_998941.1 | 5 | 949 | 100 | |
| GenBank mRNA | AB116556.1 | - | 5 | 949 | 100 | |
| AK231766.1 | - | 4 | 1023 | 100 | ||
| AK239691.1 | - | 5 | 993 | 100 | ||
| AK240451.1 | - | 5 | 998 | 100 | ||
| AK351901.1 | - | 5 | 988 | 100 | ||
| AK392860.1 | - | 5 | 1056 | 100 | ||
| AK396646.1 | - | 5 | 1011 | 100 | ||
| AK399108.1 | - | 5 | 992 | 100 | ||
More Information
Set Membership: |
|

CD2
Genomic Map