Assay Details
| Gene Symbol: FATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213797.1 | NP_998962.1 | 1 | 149 | 74 | |
| GenBank mRNA | AF345986.1 | - | 1 | 149 | 74 | |
| AK238435.1 | - | 1 | 191 | 74 | ||
| AK351278.1 | - | 1 | 166 | 74 | ||
More Information
Set Membership: |
|

FATE1
Genomic Map