Assay Details
| Gene Symbol: FGF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213801.1 | NP_998966.1 | 3 | 648 | 68 | |
| GenBank mRNA | AK343298.1 | - | 3 | 533 | 68 | |
| AK393608.1 | - | 3 | 283 | 68 | ||
| AY033825.2 | - | 3 | 648 | 68 | ||
More Information
Set Membership: |
|

FGF9
Genomic Map