Assay Details
| Gene Symbol: DEFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213838.1 | NP_999003.1 | 2 | 143 | 72 | |
| GenBank mRNA | AF031666.1 | - | 2 | 138 | 72 | |
| JQ687225.1 | - | 2 | 135 | 72 | ||
| KP862533.1 | - | 2 | 135 | 72 | ||
More Information
Set Membership: |
|

DEFB1
Genomic Map