Assay Details
| Gene Symbol: POMC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213858.1 | NP_999023.1 | 3 | 446 | 158 | |
| XM_021085834.1 | XP_020941493.1 | 665 | 158 | |||
| GenBank mRNA | AK349473.1 | - | 3 | 492 | 158 | |
| S73519.1 | - | 3 | 498 | 158 | ||
| X00135.1 | - | 498 | 158 | |||
| X03561.1 | - | 446 | 158 | |||
More Information
Set Membership: |
|

POMC
Genomic Map