Assay Details
| Gene Symbol: TNNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213912.3 | NP_999077.1 | 6 | 293 | 150 | |
| XM_021064063.1 | XP_020919722.1 | 510 | 150 | |||
| GenBank mRNA | AY282922.3 | - | 6 | 293 | 150 | |
| EU732736.1 | - | 3 | 212 | 150 | ||
| FJ719786.1 | - | 6 | 293 | 150 | ||
More Information
Set Membership: |
|

TNNI1
Genomic Map