Assay Details
| Gene Symbol: CBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214073.1 | NP_999238.1 | 3 | 855 | 89 | |
| GenBank mRNA | AK351186.1 | - | 3 | 876 | 89 | |
| AK351189.1 | - | 3 | 940 | 89 | ||
| AK398143.1 | - | 3 | 941 | 89 | ||
| AK401021.1 | - | 3 | 943 | 89 | ||
| M80709.1 | - | 3 | 855 | 89 | ||
More Information
Set Membership: |
|

CBR1
Genomic Map