Assay Details
| Gene Symbol: HTR1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214101.1 | NP_999266.1 | 1 | 953 | 137 | |
| XM_005670286.3 | XP_005670343.2 | 2729 | 137 | |||
| GenBank mRNA | AF255663.1 | - | 1 | 953 | 137 | |
More Information
Set Membership: |
|

HTR1F
Genomic Map