Assay Details
| Gene Symbol: MIOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214102.2 | NP_999267.1 | 9 | 746 | 79 | |
| XM_005653181.3 | XP_005653238.1 | 908 | 79 | |||
| GenBank mRNA | AF401311.1 | - | 9 | 750 | 79 | |
| AK232286.1 | - | 9 | 748 | 79 | ||
| AK345316.1 | - | 9 | 751 | 79 | ||
| AK346272.1 | - | 9 | 748 | 79 | ||
| AK400525.1 | - | 9 | 751 | 79 | ||
More Information
Set Membership: |
|

MIOX
Genomic Map