Assay Details
| Gene Symbol: MC4R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214173.1 | NP_999338.1 | 1 | 1453 | 135 | |
| GenBank mRNA | AB021664.1 | - | 1 | 1453 | 135 | |
| DQ388767.1 | - | 1 | 919 | 135 | ||
| JQ828977.1 | - | 1 | 959 | 135 | ||
More Information
Set Membership: |
|

MC4R
Genomic Map