Assay Details
| Gene Symbol: PPP1CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214184.2 | NP_999349.1 | 2 | 150 | 92 | |
| XM_021085856.1 | XP_020941515.1 | 444 | 92 | |||
| XM_021085857.1 | XP_020941516.1 | 347 | 92 | |||
| GenBank mRNA | AB016735.1 | - | 2 | 137 | 92 | |
| AK239060.1 | - | 3 | 343 | 92 | ||
| DQ396471.1 | - | 2 | 150 | 92 | ||
More Information
Set Membership: |
|

PPP1CB
Genomic Map