Assay Details
| Gene Symbol: DDOST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214188.1 | NP_999353.1 | 11 | 1257 | 57 | |
| GenBank mRNA | AJ293581.1 | - | 11 | 1257 | 57 | |
| AK232685.1 | - | 11 | 1216 | 57 | ||
| AK392807.1 | - | 11 | 1257 | 57 | ||
| AK392929.1 | - | 11 | 1250 | 57 | ||
More Information
Set Membership: |
|

DDOST
Genomic Map