Assay Details
| Gene Symbol: ACP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214209.1 | NP_999374.1 | 5 | 1048 | 67 | |
| XM_021081258.1 | XP_020936917.1 | 1421 | 67 | |||
| XM_021081262.1 | XP_020936921.1 | 1574 | 67 | |||
| XM_021081263.1 | XP_020936922.1 | 1099 | 67 | |||
| GenBank mRNA | AK345814.1 | - | 5 | 1063 | 67 | |
| M30284.1 | - | 5 | 1044 | 67 | ||
| M98553.1 | - | 5 | 1050 | 67 | ||
More Information
Set Membership: |
|

ACP5
Genomic Map