Assay Details
| Gene Symbol: HSD11B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348961.2 | NP_001335890.1 | 5 | 748 | 99 | |
| NM_214248.3 | NP_999413.2 | 6 | 879 | 99 | ||
| XM_021102234.1 | XP_020957893.1 | 1050 | 99 | |||
| XM_021102235.1 | XP_020957894.1 | 926 | 99 | |||
| XM_021102237.1 | XP_020957896.1 | 946 | 99 | |||
| XM_021102238.1 | XP_020957897.1 | 1035 | 99 | |||
| XM_021102239.1 | XP_020957898.1 | 909 | 99 | |||
| GenBank mRNA | AF414124.1 | - | 6 | 854 | 99 | |
| AK344628.1 | - | 6 | 775 | 99 | ||
| AK344692.1 | - | 7 | 904 | 99 | ||
| AK391330.1 | - | 6 | 771 | 99 | ||
| AK391467.1 | - | 7 | 899 | 99 | ||
| AK394626.1 | - | 7 | 916 | 99 | ||
| AK400759.1 | - | 7 | 918 | 99 | ||
| KP735788.1 | - | 6 | 954 | 99 | ||
More Information
Set Membership: |
|

HSD11B1
Genomic Map