Assay Details
| Gene Symbol: TGFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214251.1 | NP_999416.1 | 4 | 250 | 69 | |
| XM_021085860.1 | XP_020941519.1 | 433 | 69 | |||
| GenBank mRNA | AK395868.1 | - | 4 | 440 | 69 | |
| X71014.1 | - | 250 | 69 | |||
More Information
Set Membership: |
|

TGFA
Genomic Map