Assay Details
| Gene Symbol: MX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097416.1 | NP_001090885.1 | 8 | 1352 | 64 | |
| GenBank mRNA | AK345001.1 | - | 8 | 1336 | 64 | |
| AY897395.1 | - | 8 | 1352 | 64 | ||
| M65088.1 | - | 6 | 781 | 64 | ||
More Information
Set Membership: |
|

MX2
Genomic Map