Assay Details
| Gene Symbol: ITGB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097424.1 | NP_001090893.1 | 13 | 2152 | 144 | |
| XM_021101901.1 | XP_020957560.1 | 3656 | 144 | |||
| XM_021101902.1 | XP_020957561.1 | 3524 | 144 | |||
| XM_021101903.1 | XP_020957562.1 | 2577 | 144 | |||
| XM_021101904.1 | XP_020957563.1 | 2270 | 144 | |||
| GenBank mRNA | DQ786569.1 | - | 13 | 2556 | 144 | |
| EF432730.1 | - | 13 | 2152 | 144 | ||
More Information
Set Membership: |
|

ITGB8
Genomic Map